Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025604.2 | NP_001020775.1 | 8 | 1421 | ||
NM_001286826.1 | NP_001273755.1 | 8 | 1393 | |||
NM_015683.1 | NP_056498.1 | 8 | 1396 | |||
GenBank mRNA | AF131826.1 | - | 7 | 928 | ||
AF193051.1 | - | 8 | 1396 | |||
AK000689.1 | - | 8 | 1391 | |||
AK127940.1 | - | 8 | 1385 | |||
AK128588.1 | - | 2 | 2643 | |||
BC022516.1 | - | 8 | 1394 | 4817429 |