Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024948.2 | NP_001020119.1 | 4 | 409 | ||
NM_001164473.2 | NP_001157945.1 | 4 | 409 | |||
NM_017737.4 | NP_060207.2 | 4 | 409 | |||
XM_011541625.2 | XP_011539927.1 | 4 | 473 | |||
XM_017001533.1 | XP_016857022.1 | 5 | 604 | |||
GenBank mRNA | AY514449.1 | - | 4 | 258 |