Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017845.3 | NP_060315.1 | 2 | 206 | ||
XM_017008330.1 | XP_016863819.1 | 2 | 249 | |||
GenBank mRNA | AF182421.1 | - | 2 | 236 | ||
AK000509.1 | - | 2 | 226 | |||
AY542163.1 | - | 2 | 198 | |||
BC008371.1 | - | 2 | 266 | 4051246 | ||
BC015145.1 | - | 2 | 210 | 4052767 | ||
BC019826.1 | - | 2 | 232 | 4995315 |