Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290020.1 | NP_001276949.1 | 5 | 759 | ||
NM_018076.4 | NP_060546.2 | 5 | 712 | |||
XM_011519527.1 | XP_011517829.1 | 5 | 759 | |||
XM_011519528.1 | XP_011517830.1 | 5 | 1409 | |||
XM_017016368.1 | XP_016871857.1 | 5 | 759 | |||
XM_017016369.1 | XP_016871858.1 | 5 | 759 | |||
XM_017016370.1 | XP_016871859.1 | 5 | 759 | |||
XM_017016371.1 | XP_016871860.1 | 5 | 759 | |||
GenBank mRNA | AK001238.1 | - | 3 | 299 | ||
AK001679.1 | - | 5 | 710 | |||
AK292521.1 | - | 5 | 715 | |||
BC140846.1 | - | 4 | 640 | 9021714 |