Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286772.1 | NP_001273701.1 | 7 | 625 | ||
NM_018201.4 | NP_060671.3 | 7 | 625 | |||
XM_005252060.2 | XP_005252117.1 | 6 | 528 | |||
GenBank mRNA | AB209565.1 | - | 4 | 344 | ||
AB449899.1 | - | 7 | 478 | |||
AK001605.1 | - | 7 | 483 | |||
AK123828.1 | - | 3 | 239 | |||
AK289777.1 | - | 7 | 537 | |||
AK295183.1 | - | 4 | 328 | |||
AK307556.1 | - | 3 | 227 | |||
BC014898.1 | - | 7 | 500 | |||
BC132817.1 | - | 7 | 498 | 40146839 | ||
BC136509.1 | - | 7 | 498 | 9020499 | ||
BC150311.1 | - | 7 | 534 | 8860447 |