Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020801.2 | NP_065852.1 | 5 | 983 | ||
GenBank mRNA | AB037797.1 | - | 5 | 955 | ||
AK291753.1 | - | 5 | 958 | |||
BC015928.2 | - | 5 | 950 | 3908916 | ||
BC053619.1 | - | 5 | 955 | 6155815 |