Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024808.2 | NP_001019979.1 | 3 | 415 | ||
NM_020993.4 | NP_066273.1 | 3 | 415 | |||
GenBank mRNA | AK296159.1 | - | 3 | 417 | ||
AK316486.1 | - | 3 | 313 | |||
BC094723.1 | - | 3 | 226 | 5300018 | ||
BC113627.1 | - | 3 | 239 | 8322679 | ||
BM801030.1 | - | 374 | ||||
X89984.1 | - | 3 | 1162 |