Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021185.4 | NP_067008.3 | 27 | 3191 | ||
XM_005259114.4 | XP_005259171.1 | 27 | 3287 | |||
XM_005259115.4 | XP_005259172.1 | 27 | 3210 | |||
XM_005259116.2 | XP_005259173.1 | 25 | 3011 | |||
XM_006723308.3 | XP_006723371.1 | 27 | 3807 | |||
XM_006723310.2 | XP_006723373.1 | 27 | 3074 | |||
XM_006723311.2 | XP_006723374.1 | 27 | 3212 | |||
XM_011527174.2 | XP_011525476.1 | 27 | 3201 | |||
XM_011527175.2 | XP_011525477.1 | 27 | 3107 | |||
XM_011527176.2 | XP_011525478.1 | 27 | 3110 | |||
XM_011527177.2 | XP_011525479.1 | 27 | 3071 | |||
XM_011527178.2 | XP_011525480.1 | 28 | 3553 | |||
XM_011527182.2 | XP_011525484.1 | 26 | 2935 | |||
XM_017027053.1 | XP_016882542.1 | 26 | 3071 | |||
XM_017027054.1 | XP_016882543.1 | 25 | 2804 | |||
XM_017027055.1 | XP_016882544.1 | 14 | 2280 | |||
GenBank mRNA | AK096218.1 | - | 2 | 1349 | ||
AK128220.1 | - | 27 | 2974 | |||
AK310460.1 | - | 24 | 2700 | |||
BC109035.1 | - | 23 | 2590 | 40028284 |