Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024761.4 | NP_079037.3 | 4 | 5794 | ||
GenBank mRNA | AA149400.1 | - | 1 | 100 | ||
CB128567.1 | - | 1 | 220 |