Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289119.1 | NP_001276048.1 | 9 | 2658 | ||
NM_001289120.1 | NP_001276049.1 | 9 | 2681 | |||
NM_001289121.1 | NP_001276050.1 | 9 | 2799 | |||
NM_032226.2 | NP_115602.2 | 9 | 2654 | |||
NR_110317.1 | 8 | 2023 | ||||
XM_005251608.4 | XP_005251665.1 | 9 | 3563 | |||
XM_005251612.2 | XP_005251669.1 | 9 | 2621 | |||
XM_011518050.1 | XP_011516352.1 | 8 | 2553 | |||
XM_011518051.2 | XP_011516353.1 | 8 | 7586 | |||
XM_017015197.1 | XP_016870686.1 | 8 | 2680 | |||
XM_017015198.1 | XP_016870687.1 | 8 | 3166 | |||
XM_017015199.1 | XP_016870688.1 | 8 | 2579 | |||
XM_017015200.1 | XP_016870689.1 | 8 | 2852 | |||
XM_017015201.1 | XP_016870690.1 | 8 | 1998 | |||
GenBank mRNA | AK074608.1 | - | 8 | 2143 | ||
BC036940.1 | - | 9 | 2772 | 5399765 |