Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AFAP1L2 SIRNA Assays ›
Gene Symbol:
AFAP1L2
Gene Name:
actin filament associated protein 1 like 2
Gene Aliases:
CTB-1144G6.4, KIAA1914, XB130
Chromosome Location:
Chr.10: 114280730 - 114405143 on Build GRCh38
UniGene ID:
Hs.501106
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147942
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AFAP1L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001936.2
NP_001001936.1
16
2296
NM_001287824.1
NP_001274753.1
17
2455
NM_032550.3
NP_115939.1
16
2296
XM_005270233.4
XP_005270290.1
17
2244
XM_005270237.4
XP_005270294.1
16
2020
XM_005270241.4
XP_005270298.1
17
2255
XM_005270242.4
XP_005270299.1
14
1842
XM_011540286.2
XP_011538588.1
16
2066
XM_011540289.1
XP_011538591.1
16
2066
XM_011540290.1
XP_011538592.1
16
2066
XM_017016792.1
XP_016872281.1
17
2150
XM_017016793.1
XP_016872282.1
17
2147
XM_017016794.1
XP_016872283.1
17
2150
XM_017016795.1
XP_016872284.1
18
2328
XM_017016796.1
XP_016872285.1
17
2102
XM_017016797.1
XP_016872286.1
17
2099
XM_017016798.1
XP_016872287.1
17
2102
XM_017016799.1
XP_016872288.1
17
2150
XM_017016800.1
XP_016872289.1
17
2150
XM_017016801.1
XP_016872290.1
17
2147
XM_017016802.1
XP_016872291.1
17
2150
XM_017016803.1
XP_016872292.1
17
2147
XM_017016804.1
XP_016872293.1
16
2020
XM_017016805.1
XP_016872294.1
17
2102
XM_017016806.1
XP_016872295.1
17
2099
XM_017016807.1
XP_016872296.1
16
2063
XM_017016808.1
XP_016872297.1
17
2102
XM_017016809.1
XP_016872298.1
17
2099
XM_017016810.1
XP_016872299.1
17
2244
XM_017016811.1
XP_016872300.1
17
2241
XM_017016812.1
XP_016872301.1
17
2244
XM_017016813.1
XP_016872302.1
17
2241
XM_017016814.1
XP_016872303.1
18
2339
XM_017016815.1
XP_016872304.1
16
1901
XM_017016816.1
XP_016872305.1
16
2017
XM_017016817.1
XP_016872306.1
16
2017
XM_017016818.1
XP_016872307.1
16
1853
XM_017016819.1
XP_016872308.1
16
1901
XM_017016820.1
XP_016872309.1
18
2339
XM_017016821.1
XP_016872310.1
18
2336
XM_017016822.1
XP_016872311.1
16
1901
XM_017016823.1
XP_016872312.1
16
1853
XM_017016824.1
XP_016872313.1
16
1853
XM_017016825.1
XP_016872314.1
17
2255
XM_017016826.1
XP_016872315.1
17
2252
XM_017016827.1
XP_016872316.1
15
1771
XM_017016828.1
XP_016872317.1
15
1771
XM_017016829.1
XP_016872318.1
15
1924
XM_017016830.1
XP_016872319.1
16
2006
XM_017016831.1
XP_016872320.1
15
1921
XM_017016832.1
XP_016872321.1
14
1884
GenBank mRNA
AB067501.1
-
4
1095
AF474151.1
-
16
2022
AK123108.1
-
17
2066
AK291712.1
-
16
2020
AK294969.1
-
17
2181
AK295538.1
-
6
917
BC024314.1
-
16
2274
3900921
BC033212.1
-
16
2003
4909140
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
regulation of mitotic cell cycle
regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of protein tyrosine kinase activity
SH3 domain binding
protein tyrosine kinase activator activity
signaling adaptor activity
SH2 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0