Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164097.1 | NP_001157569.1 | 7 | 3754 | ||
NM_004385.4 | NP_004376.2 | 8 | 6715 | |||
GenBank mRNA | AB209491.1 | - | 8 | 6598 | ||
AK299770.1 | - | 3 | 2672 | |||
AK307642.1 | - | 7 | 3655 | |||
BX641036.1 | - | 1 | 3749 | |||
U16306.1 | - | 8 | 6625 | |||
X15998.1 | - | 7 | 3664 |