Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018037.1 | NP_001018047.1 | 4 | 500 | ||
NM_001018038.2 | NP_001018048.1 | 4 | 500 | |||
NM_015186.3 | NP_056001.1 | 4 | 500 | |||
NM_033305.2 | NP_150648.2 | 4 | 500 | |||
GenBank mRNA | AB054005.1 | - | 4 | 411 | ||
AF337532.1 | - | 4 | 500 | |||
AJ608769.1 | - | 4 | 500 | |||
AJ626859.1 | - | 4 | 500 |