Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300922.1 | NP_001287851.1 | 3 | 268 | ||
NM_054113.3 | NP_473454.1 | 5 | 415 | |||
GenBank mRNA | AB050868.1 | - | 5 | 365 | ||
BC069320.1 | - | 3 | 105 | |||
BC069428.1 | - | 3 | 218 | 7262131 | ||
BC069524.1 | - | 5 | 346 | |||
BC112200.1 | - | 5 | 415 | 8327668 | ||
BC113591.1 | - | 5 | 415 | 8322643 |