Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139073.3 | NP_620712.2 | 1 | 286 | ||
XM_005246303.2 | XP_005246360.1 | 1 | 284 | |||
XM_017003363.1 | XP_016858852.1 | 1 | 284 | |||
GenBank mRNA | AK093385.1 | - | 1 | 284 | ||
AK126086.1 | - | 2 | 232 | |||
AY032925.1 | - | 2 | 384 | |||
BC047704.1 | - | 2 | 229 | 5744971 | ||
DB034232.1 | - | 1 | 284 |