Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004415.3 | NP_004406.2 | 23 | 4780 | ||
GenBank mRNA | J05211.1 | - | 1 | 1082 | ||
M77830.3 | - | 23 | 4718 |