Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317968.1 | NP_001304897.1 | 4 | 466 | ||
NM_152285.3 | NP_689498.1 | 4 | 466 | |||
XM_005266119.1 | XP_005266176.1 | 4 | 465 | |||
XM_006717320.3 | XP_006717383.1 | 3 | 837 | |||
XM_006717321.3 | XP_006717384.1 | 3 | 837 | |||
XM_006717322.3 | XP_006717385.1 | 3 | 837 | |||
XM_017015288.1 | XP_016870777.1 | 3 | 837 | |||
GenBank mRNA | AB209523.1 | - | 1 | 917 | ||
AJ420420.1 | - | 4 | 429 | |||
AK001822.1 | - | 3 | 726 | |||
AK308376.1 | - | 4 | 403 | |||
AL542109.3 | - | 4 | 462 | |||
BC032346.1 | - | 4 | 452 | 5211669 | ||
BI906381.1 | - | 4 | 467 | |||
CR627251.1 | - | 3 | 5402 |