Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152484.2 | NP_689697.2 | 6 | 2894 | ||
XM_006723046.2 | XP_006723109.1 | 8 | 3221 | |||
XM_006723047.3 | XP_006723110.1 | 7 | 3144 | |||
XM_006723048.3 | XP_006723111.1 | 7 | 3068 | |||
XM_011526538.2 | XP_011524840.1 | 7 | 2983 | |||
XM_011526539.2 | XP_011524841.1 | 5 | 2477 | |||
XM_017026376.1 | XP_016881865.1 | 6 | 2729 | |||
XM_017026377.1 | XP_016881866.1 | 6 | 2808 | |||
XM_017026378.1 | XP_016881867.1 | 7 | 2846 | |||
XM_017026379.1 | XP_016881868.1 | 4 | 2695 | |||
XM_017026380.1 | XP_016881869.1 | 4 | 2503 | |||
XM_017026381.1 | XP_016881870.1 | 5 | 2906 | |||
XM_017026382.1 | XP_016881871.1 | 4 | 2603 | |||
GenBank mRNA | AK056615.1 | - | 1 | 1691 | ||
AK122846.1 | - | 3 | 2215 | |||
AK289987.1 | - | 6 | 2667 | |||
AL833408.1 | - | 6 | 2895 | |||
AY835704.1 | - | 6 | 2894 | |||
BC059407.1 | - | 5 | 2523 |