Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304963.1 | NP_001291892.1 | 2 | 1829 | ||
NM_153183.3 | NP_694853.1 | 3 | 1892 | |||
GenBank mRNA | AF469196.1 | - | 3 | 1894 | ||
BC049383.1 | - | 2 | 1750 | 5725355 | ||
BC050700.1 | - | 2 | 1808 | 5723622 |