Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BDNF SIRNA Assays ›
Gene Symbol:
BDNF
Gene Name:
brain derived neurotrophic factor
Gene Aliases:
ANON2, BULN2
Chromosome Location:
Chr.11: 27654893 - 27722058 on Build GRCh38
UniGene ID:
Hs.502182
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148177
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BDNF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143805.1
NP_001137277.1
2
2021
NM_001143806.1
NP_001137278.1
2
2236
NM_001143807.1
NP_001137279.1
2
2122
NM_001143808.1
NP_001137280.1
2
1958
NM_001143809.1
NP_001137281.1
2
1968
NM_001143810.1
NP_001137282.1
3
2251
NM_001143811.1
NP_001137283.1
4
2368
NM_001143812.1
NP_001137284.1
2
2110
NM_001143813.1
NP_001137285.1
2
2192
NM_001143814.1
NP_001137286.1
3
2317
NM_001143816.1
NP_001137288.1
2
2715
NM_001709.4
NP_001700.2
2
2210
NM_170731.4
NP_733927.1
2
2532
NM_170732.4
NP_733928.1
2
2319
NM_170733.3
NP_733929.1
2
2222
NM_170734.3
NP_733930.1
2
2170
NM_170735.5
NP_733931.1
1
2949
XM_011520280.2
XP_011518582.1
2
3117
GenBank mRNA
AY054392.2
-
2
2223
AY054393.2
-
2
1966
AY054394.2
-
2
2180
AY054395.2
-
2
2263
AY054396.2
-
2
2021
AY054397.2
-
2
2104
AY054398.2
-
2
2151
AY054399.2
-
3
2036
AY054400.2
-
2
2137
EF674517.1
-
2
2021
EF674518.1
-
2
2236
EF674519.1
-
2
2319
EF674520.1
-
2
2122
EF674521.1
-
2
2222
EF689009.1
-
1
2963
EF689010.1
-
2
2729
EF689011.1
-
2
1958
EF689012.1
-
2
1968
EF689013.1
-
2
2110
EF689014.1
-
2
2192
EF689015.1
-
2
2210
EF689016.1
-
3
2317
EF689017.1
-
3
2069
EF689018.1
-
2
2170
EF689019.1
-
3
2251
EF689020.1
-
4
2368
EF689021.1
-
2
2532
Back To Top
More Information
Panther Classification:
Molecular Function -
neurotrophic factor
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell signaling
nervous system development
axon guidance
synapse assembly
brain-derived neurotrophic factor receptor signaling pathway
positive regulation of brain-derived neurotrophic factor receptor signaling pathway
negative regulation of neuron apoptotic process
collateral sprouting
positive regulation of collateral sprouting
positive regulation of synapse assembly
neurotrophin TRKB receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0