Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM117B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173511.3 | NP_775782.2 | 8 | 2405 | ||
GenBank mRNA | AK096090.1 | - | 8 | 1993 | ||
BC065010.1 | - | 7 | 1805 | |||
BX537704.1 | - | 1 | 1021 |