Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGEF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175062.3 | NP_778232.2 | 14 | 1851 | ||
GenBank mRNA | AF449764.1 | - | 13 | 2037 | ||
AK093160.1 | - | 11 | 1702 | |||
BC036802.1 | - | 15 | 1868 | |||
BC057759.1 | - | 12 | 1542 | 5268021 |