Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201628.2 | NP_963922.2 | 15 | 2684 | ||
XM_005245795.4 | XP_005245852.1 | 17 | 2939 | |||
XM_011541074.2 | XP_011539376.1 | 16 | 4875 | |||
XM_011541075.2 | XP_011539377.1 | 14 | 4531 | |||
XM_011541076.2 | XP_011539378.1 | 14 | 4676 | |||
XM_011541077.2 | XP_011539379.1 | 14 | 5655 | |||
XM_011541081.2 | XP_011539383.1 | 8 | 3355 | |||
GenBank mRNA | AK293806.1 | - | 10 | 1490 |