Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201628.2 | NP_963922.2 | 15 | 3792 | ||
XM_005245795.4 | XP_005245852.1 | 17 | 4047 | |||
XM_011541074.2 | XP_011539376.1 | 16 | 5983 | |||
XM_011541075.2 | XP_011539377.1 | 14 | 5639 | |||
XM_011541076.2 | XP_011539378.1 | 14 | 5784 | |||
XM_011541077.2 | XP_011539379.1 | 14 | 6763 | |||
XM_011541081.2 | XP_011539383.1 | 8 | 4463 | |||
GenBank mRNA | AK057155.1 | - | 1 | 352 | ||
AK125388.1 | - | 1 | 967 |