Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006346.2 | NP_006337.2 | 15 | 2193 | ||
XM_011534881.2 | XP_011533183.1 | 16 | 2308 | |||
XM_011534885.2 | XP_011533187.1 | 15 | 1935 | |||
XM_017020350.1 | XP_016875839.1 | 14 | 1847 | |||
XM_017020352.1 | XP_016875841.1 | 16 | 2257 | |||
XM_017020353.1 | XP_016875842.1 | 15 | 2167 | |||
XM_017020354.1 | XP_016875843.1 | 14 | 1962 | |||
GenBank mRNA | AF330046.1 | - | 15 | 2235 | ||
AY375528.1 | - | 6 | 651 | |||
BC051911.1 | - | 15 | 2123 | 6140514 |