Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164595.1 | NP_001158067.1 | 4 | 959 | ||
NM_013377.3 | NP_037509.3 | 2 | 383 | |||
GenBank mRNA | AK091096.1 | - | 2 | 350 | ||
AK131554.1 | - | 4 | 559 | |||
AK294431.1 | - | 2 | 439 | |||
AK294535.1 | - | 4 | 422 | |||
BC093744.1 | - | 2 | 232 | 7939589 | ||
BC093746.2 | - | 2 | 232 | 7939591 |