Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301015.1 | NP_001287944.1 | 8, 9 | 1437 | ||
NM_001301018.1 | NP_001287947.1 | 8, 9 | 911 | |||
NM_001301027.1 | NP_001287956.1 | 9, 10 | 1516 | |||
NM_001301028.1 | NP_001287957.1 | 8, 9 | 1437 | |||
NM_014945.3 | NP_055760.1 | 9, 10 | 1083 | |||
XM_005268392.1 | XP_005268449.1 | 9, 10 | 1064 | |||
XM_005268393.1 | XP_005268450.1 | 9, 10 | 1064 | |||
XM_005268394.1 | XP_005268451.1 | 9, 10 | 1064 | |||
XM_005268395.1 | XP_005268452.1 | 9, 10 | 1064 | |||
XM_005268396.1 | XP_005268453.1 | 9, 10 | 1064 | |||
XM_011537609.2 | XP_011535911.1 | 9, 10 | 1064 | |||
XM_017009238.1 | XP_016864727.1 | 8, 9 | 1158 | |||
GenBank mRNA | AB020650.1 | - | 9, 10 | 996 | ||
AK225769.1 | - | 8, 9 | 1305 | |||
AK289736.1 | - | 8, 9 | 850 | |||
AL833021.1 | - | 5, 6 | 476 | |||
BC001665.2 | - | 8, 9 | 1099 | 2988043 | ||
DQ413174.1 | - | 8, 9 | 873 |