Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014974.2 | NP_055789.1 | 37 | 5300 | ||
XM_005252426.3 | XP_005252483.1 | 38 | 5459 | |||
XM_005252427.3 | XP_005252484.1 | 38 | 5370 | |||
XM_005252428.4 | XP_005252485.1 | 39 | 5540 | |||
XM_005252430.2 | XP_005252487.2 | 21 | 3550 | |||
XM_011519428.2 | XP_011517730.1 | 39 | 5471 | |||
XM_011519429.2 | XP_011517731.1 | 38 | 5439 | |||
XM_011519430.2 | XP_011517732.1 | 38 | 5346 | |||
XM_011519431.2 | XP_011517733.1 | 37 | 5316 | |||
XM_011519432.1 | XP_011517734.1 | 38 | 5771 | |||
XM_011519433.2 | XP_011517735.1 | 22 | 4579 | |||
XM_017015977.1 | XP_016871466.1 | 22 | 3591 | |||
GenBank mRNA | AB023151.2 | - | 37 | 5299 | ||
AK311679.1 | - | 2 | 820 | |||
BX537672.1 | - | 1 | 2657 |