Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297669.1 | NP_001284598.1 | 19 | 3320 | ||
NM_001297670.1 | NP_001284599.1 | 18 | 3210 | |||
NM_001297671.1 | NP_001284600.1 | 18 | 2946 | |||
NM_001297672.1 | NP_001284601.1 | 17 | 2859 | |||
NM_015149.4 | NP_055964.3 | 19 | 3321 | |||
XM_011509339.2 | XP_011507641.1 | 20 | 3367 | |||
XM_011509341.1 | XP_011507643.1 | 18 | 3019 | |||
XM_017000756.1 | XP_016856245.1 | 17 | 2540 | |||
GenBank mRNA | AB023176.1 | - | 18 | 2886 | ||
AF186779.1 | - | 18 | 2907 | |||
AF186780.1 | - | 19 | 3283 | |||
AL080117.1 | - | 17 | 2689 | |||
BC136591.1 | - | 19 | 3025 | 9020581 | ||
BX537416.1 | - | 19 | 3261 |