Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004525.2 | NP_004516.2 | 64 | 12114 | ||
XM_011511183.2 | XP_011509485.1 | 63 | 11986 | |||
XM_011511184.2 | XP_011509486.1 | 49 | 9627 | |||
GenBank mRNA | AK302843.1 | - | 15 | 2013 | ||
AY265357.1 | - | 64 | 11968 | |||
AY265358.1 | - | 64 | 11965 | |||
U04441.1 | - | 22 | 3795 | |||
U33837.1 | - | 64 | 12005 |