Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015329.3 | NP_056144.3 | 19 | 2210 | ||
XM_005259837.3 | XP_005259894.2 | 19 | 2199 | |||
XM_005259838.3 | XP_005259895.3 | 17 | 2261 | |||
XM_011527844.2 | XP_011526146.1 | 19 | 2615 | |||
XM_017026539.1 | XP_016882028.1 | 17 | 2257 | |||
GenBank mRNA | AB020699.1 | - | 19 | 2196 | ||
AK001349.1 | - | 7 | 927 | |||
AK025285.1 | - | 1 | 845 | |||
AK025896.1 | - | 17 | 1862 | |||
AK098275.1 | - | 2 | 961 | |||
AK125914.1 | - | 4 | 2429 | |||
AK126227.1 | - | 6 | 2102 | |||
AK128029.1 | - | 10 | 2194 | |||
AL110250.1 | - | 8 | 985 | |||
BC010222.1 | - | 8 | 996 | |||
BC063863.1 | - | 8 | 1343 |