Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS13D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015378.3 | NP_056193.2 | 13 | 1712 | ||
NM_018156.3 | NP_060626.2 | 13 | 1712 | |||
GenBank mRNA | AJ608774.1 | - | 13 | 1684 | ||
AJ608775.1 | - | 13 | 1684 | |||
AL162331.1 | - | 13 | 1684 |