Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015483.2 | NP_056298.2 | 4 | 1180 | ||
XM_005249696.2 | XP_005249753.1 | 4 | 1036 | |||
XM_005249697.2 | XP_005249754.1 | 4 | 769 | |||
XM_011515256.2 | XP_011513558.1 | 4 | 915 | |||
XM_011515257.2 | XP_011513559.1 | 4 | 649 | |||
GenBank mRNA | AB040922.1 | - | 1 | 1804 | ||
AK024557.1 | - | 1 | 145 | |||
AK292802.1 | - | 4 | 902 | |||
BC037887.2 | - | 4 | 1084 | 5274706 | ||
BC047107.1 | - | 4 | 917 | 5275217 |