Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004540.3 | NP_004531.2 | 17 | 2582 | ||
XM_011529575.2 | XP_011527877.1 | 18 | 3057 | |||
XM_011529576.2 | XP_011527878.1 | 18 | 2634 | |||
XM_011529579.2 | XP_011527881.1 | 17 | 2558 | |||
XM_011529585.2 | XP_011527887.1 | 14 | 1924 | |||
XM_017028353.1 | XP_016883842.1 | 18 | 2722 | |||
XM_017028356.1 | XP_016883845.1 | 17 | 2457 | |||
XM_017028357.1 | XP_016883846.1 | 17 | 2457 | |||
GenBank mRNA | AK299054.1 | - | 14 | 2095 | ||
BC052946.1 | - | 17 | 2582 | 5315109 | ||
U75330.1 | - | 17 | 2402 |