Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004541.3 | NP_004532.1 | 2, 3 | 341 | ||
GenBank mRNA | AB451304.1 | - | 2, 3 | 200 | ||
AB451435.1 | - | 2, 3 | 200 | |||
BC000266.1 | - | 2, 3 | 238 | 3352028 | ||
BQ680674.1 | - | 2, 3 | 242 | |||
CR456889.1 | - | 2, 3 | 200 | |||
U54993.1 | - | 2, 3 | 233 | |||
X81900.1 | - | 2, 3 | 217 |