Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016094.3 | NP_057178.2 | 3 | 260 | ||
NR_135149.1 | 3 | 260 | ||||
GenBank mRNA | AF059618.1 | - | 3 | 236 | ||
AY542158.1 | - | 3 | 205 | |||
BC001228.1 | - | 3 | 239 | |||
BC022206.1 | - | 3 | 244 | |||
BC046131.1 | - | 3 | 222 | 4443942 | ||
BC093077.1 | - | 3 | 250 | 30372667 | ||
BG700015.1 | - | 3 | 262 | |||
CN483677.1 | - | 3 | 211 | |||
DA811158.1 | - | 3 | 259 |