Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286703.1 | NP_001273632.1 | 6 | 1035 | ||
NM_001286704.1 | NP_001273633.1 | 6 | 1213 | |||
NM_001286705.1 | NP_001273634.1 | 7 | 1077 | |||
NM_001286706.1 | NP_001273635.1 | 6 | 1711 | |||
NM_016617.3 | NP_057701.1 | 6 | 1041 | |||
NR_104584.1 | 7 | 1102 | ||||
NR_104585.1 | 5 | 1001 | ||||
GenBank mRNA | AF208844.1 | - | 6 | 931 | ||
AK095490.1 | - | 6 | 961 | |||
AL515797.3 | - | 5 | 893 | |||
CN362375.1 | - | 1 | 55 |