Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWF19L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303404.1 | NP_001290333.1 | 6 | 653 | ||
NM_001303405.1 | NP_001290334.1 | 6 | 698 | |||
NM_001303406.1 | NP_001290335.1 | 3 | 387 | |||
NM_001303407.1 | NP_001290336.1 | 5 | 422 | |||
NM_018294.5 | NP_060764.3 | 6 | 653 | |||
GenBank mRNA | AK001860.1 | - | 6 | 599 | ||
AK023984.1 | - | 4 | 255 | |||
AK055313.1 | - | 1 | 1140 | |||
AK097895.1 | - | 5 | 421 | |||
AK225590.1 | - | 6 | 611 | |||
AK295303.1 | - | 3 | 357 | |||
AL832515.1 | - | 6 | 610 | |||
BC008746.2 | - | 6 | 594 | 3345869 | ||
KU178692.1 | - | 6 | 565 | |||
KU178693.1 | - | 6 | 549 |