Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321772.1 | NP_001308701.1 | 3 | 1921 | ||
NM_020317.4 | NP_064713.3 | 3 | 917 | |||
NR_135143.1 | 3 | 917 | ||||
NR_135144.1 | 3 | 917 | ||||
NR_135777.1 | 3 | 892 | ||||
NR_135778.1 | 3 | 917 | ||||
NR_135780.1 | 3 | 917 | ||||
NR_135781.1 | 3 | 917 | ||||
NR_135782.1 | 3 | 917 | ||||
NR_135783.1 | 3 | 917 | ||||
NR_135784.1 | 3 | 917 | ||||
NR_135785.1 | 3 | 917 | ||||
NR_135786.1 | 3 | 917 | ||||
NR_135787.1 | 3 | 1921 | ||||
NR_135788.1 | 3 | 983 | ||||
NR_135789.1 | 3 | 1921 | ||||
XM_011541797.1 | XP_011540099.1 | 3 | 882 | |||
GenBank mRNA | AF247168.1 | - | 3 | 844 | ||
AF267856.1 | - | 3 | 843 | |||
AK027318.1 | - | 3 | 852 | |||
AK226012.1 | - | 3 | 855 | |||
AK292532.1 | - | 3 | 852 | |||
AK296798.1 | - | 4 | 648 | |||
BC001892.1 | - | 3 | 841 | 3534629 | ||
BC041843.1 | - | 3 | 1832 | 5269181 | ||
BC065040.1 | - | 3 | 830 | 6147639 |