Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020762.2 | NP_065813.1 | 22 | 4014 | ||
XM_011538580.1 | XP_011536882.1 | 23 | 4122 | |||
XM_011538581.2 | XP_011536883.1 | 23 | 5627 | |||
GenBank mRNA | AB037725.1 | - | 21 | 5236 | ||
AK023899.1 | - | 12 | 2744 | |||
AK025819.1 | - | 2 | 1252 | |||
BC053903.1 | - | 22 | 4014 | 6526787 |