Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024335.2 | NP_077311.2 | 3 | 905 | ||
XM_005256137.2 | XP_005256194.1 | 3 | 1502 | |||
GenBank mRNA | AK125053.1 | - | 3 | 1704 | ||
AK309149.1 | - | 4 | 598 | |||
AY335937.1 | - | 3 | 905 | |||
BC136573.1 | - | 3 | 646 | 9020563 | ||
BC136576.1 | - | 3 | 646 | 9020566 |