Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282658.1 | NP_001269587.1 | 7 | 2088 | ||
NM_031455.3 | NP_113643.1 | 3 | 1523 | |||
GenBank mRNA | AF190154.1 | - | 1 | 88 | ||
AK024578.1 | - | 1 | 506 | |||
AK095792.1 | - | 7 | 1967 | |||
AK123393.1 | - | 1 | 1664 | |||
AK124195.1 | - | 1 | 1226 | |||
AL136562.1 | - | 3 | 1439 | |||
BC051334.1 | - | 3 | 1523 | 6302570 |