Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESYT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322831.1 | NP_001309760.1 | 4 | 753 | ||
NM_001322834.1 | NP_001309763.1 | 4 | 753 | |||
NM_031913.4 | NP_114119.2 | 4 | 753 | |||
NR_136409.1 | 4 | 753 | ||||
XM_017007307.1 | XP_016862796.1 | 3 | 205 | |||
GenBank mRNA | AK125080.1 | - | 4 | 713 | ||
AK289530.1 | - | 4 | 710 | |||
BC037292.1 | - | 4 | 745 | 4837811 | ||
DQ993202.1 | - | 4 | 527 |