Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031949.4 | NP_114155.4 | 3 | 351 | ||
XM_006715572.3 | XP_006715635.1 | 2 | 146 | |||
GenBank mRNA | AK093039.1 | - | 3 | 351 | ||
AK225984.1 | - | 3 | 351 | |||
AK301755.1 | - | 4 | 208 | |||
AK314450.1 | - | 3 | 328 | |||
AY026506.1 | - | 3 | 323 | |||
BC030650.1 | - | 3 | 341 | 5269216 | ||
BC047411.1 | - | 3 | 329 | 5296230 |