Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168299.1 | NP_001161771.1 | 3 | 264 | ||
NM_001168300.1 | NP_001161772.1 | 2 | 512 | |||
NM_001168301.1 | NP_001161773.1 | 3 | 162 | |||
NM_001168302.1 | NP_001161774.1 | 3 | 456 | |||
NM_001168303.1 | NP_001161775.1 | 2 | 382 | |||
NM_033495.3 | NP_277030.2 | 2 | 1032 | |||
XM_011531409.2 | XP_011529711.1 | 4 | 924 | |||
XM_011531410.2 | XP_011529712.1 | 4 | 224 | |||
XM_011531411.1 | XP_011529713.1 | 3 | 603 | |||
XM_017029950.1 | XP_016885439.1 | 2 | 1293 | |||
GenBank mRNA | AB037730.1 | - | 1 | 2391 | ||
AK122724.1 | - | 2 | 99 | |||
AK125356.1 | - | 2 | 1032 | |||
AK296324.1 | - | 3 | 171 | |||
AK297523.1 | - | 2 | 204 | |||
AK299257.1 | - | 2 | 276 | |||
AK302713.1 | - | 3 | 162 | |||
AK316509.1 | - | 3 | 416 | |||
AL591986.1 | - | 2 | 179 | |||
BC064576.1 | - | 2 | 184 | 5727768 | ||
DB074659.1 | - | 2 | 383 | |||
DC343916.1 | - | 3 | 392 | |||
DC354391.1 | - | 3 | 264 |