Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168299.1 | NP_001161771.1 | 6 | 776 | ||
NM_001168300.1 | NP_001161772.1 | 5 | 1024 | |||
NM_001168301.1 | NP_001161773.1 | 6 | 674 | |||
NM_001168302.1 | NP_001161774.1 | 6 | 968 | |||
NM_001168303.1 | NP_001161775.1 | 5 | 894 | |||
NM_033495.3 | NP_277030.2 | 5 | 1544 | |||
XM_011531409.2 | XP_011529711.1 | 7 | 1436 | |||
XM_011531410.2 | XP_011529712.1 | 7 | 736 | |||
XM_011531411.1 | XP_011529713.1 | 6 | 1115 | |||
XM_017029950.1 | XP_016885439.1 | 5 | 1805 | |||
GenBank mRNA | AB037730.1 | - | 4 | 2903 | ||
AK122724.1 | - | 5 | 611 | |||
AK125356.1 | - | 5 | 1544 | |||
AK296324.1 | - | 6 | 683 | |||
AK297523.1 | - | 5 | 621 | |||
AK299257.1 | - | 5 | 788 | |||
AK302713.1 | - | 6 | 674 | |||
AK316509.1 | - | 6 | 928 | |||
AL591986.1 | - | 5 | 691 | |||
BC064576.1 | - | 5 | 696 | 5727768 |