Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARVELD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052858.5 | NP_443090.4 | 3 | 1234 | ||
GenBank mRNA | BC011893.2 | - | 3 | 1201 | 3942185 | |
BC013376.2 | - | 3 | 1201 | 3946889 |