Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301627.1 | NP_001288556.1 | 10 | 1603 | ||
NM_052917.3 | NP_443149.2 | 12 | 2081 | |||
XM_011510537.2 | XP_011508839.1 | 12 | 2076 | |||
XM_011510538.2 | XP_011508840.1 | 10 | 1488 | |||
XM_017003258.1 | XP_016858747.1 | 12 | 1930 | |||
XM_017003259.1 | XP_016858748.1 | 11 | 1655 | |||
XM_017003260.1 | XP_016858749.1 | 12 | 2076 | |||
XM_017003261.1 | XP_016858750.1 | 12 | 1940 | |||
XM_017003262.1 | XP_016858751.1 | 10 | 1346 | |||
XM_017003263.1 | XP_016858752.1 | 10 | 1488 | |||
GenBank mRNA | AB067505.1 | - | 10 | 1381 | ||
AB078142.1 | - | 10 | 1499 | |||
AJ505991.2 | - | 10 | 1499 | |||
AK123152.1 | - | 1702 | ||||
AK131195.1 | - | 12 | 2026 | |||
BC101031.1 | - | 10 | 1499 | 40007710 | ||
BC101032.2 | - | 10 | 1499 | 40007711 | ||
BC101033.1 | - | 10 | 1499 | 40007712 | ||
BC101034.2 | - | 10 | 1499 | 40007713 | ||
BG221433.1 | - | 1 | 144 | |||
KJ534843.1 | - | 10 | 1499 | |||
KJ535058.1 | - | 9 | 1330 |