Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319164.1 | NP_001306093.1 | 3 | 251 | ||
NM_080574.3 | NP_542141.1 | 3 | 333 | |||
GenBank mRNA | AF432917.1 | - | 2 | 185 | ||
AK291854.1 | - | 3 | 333 | |||
AY359055.1 | - | 3 | 251 | |||
BC065726.1 | - | 3 | 260 | 30326122 |