Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305163.1 | NP_001292092.1 | 3 | 1287 | ||
NM_001305164.1 | NP_001292093.1 | 3 | 666 | |||
NM_133368.2 | NP_588609.1 | 3 | 688 | |||
XM_005256220.1 | XP_005256277.1 | 3 | 995 | |||
XM_011523427.1 | XP_011521729.1 | 4 | 774 | |||
XM_011523428.1 | XP_011521730.1 | 4 | 1088 | |||
XM_011523430.1 | XP_011521732.1 | 4 | 715 | |||
XM_017023844.1 | XP_016879333.1 | 4 | 1461 | |||
GenBank mRNA | AB075852.1 | - | 3 | 608 | ||
AK027549.1 | - | 3 | 1238 | |||
AK172845.1 | - | 3 | 644 | |||
BC013173.2 | - | 3 | 643 | 4340287 | ||
DB083249.1 | - | 4 | 405 |